Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

691113

Reducer, SS with Ferrules 3/8" to 1/8"

Synonym(s):

Reducer for ReCycler® Bulk solvent containers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
27121703
Pricing and availability is not currently available.

material

plastic packaging
stainless steel

packaging

pkg of 1 unit

storage condition

room temperature

compatibility

for use with returnable containers

shipped in

ambient

General description

Our stainless-steel Reducer with ferrules is designed to seamlessly transition from 3/8" to 1/8" diameters, with optimal performance in ReCycler® Bulk Returnable containers.
The inclusion of ferrules facilitates a secure and leak-free connection. The design adheres to stringent industry standards and enhances reliability.

Legal Information

RECYCLER is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service