Key Documents
58258
Stainless Steel HPLC Fittings
ferrule for 1/16 in. tubing, pkg of 10 ea
About This Item
UNSPSC Code:
41115700
material
stainless steel ferrule
packaging
pkg of 10 ea
manufacturer/tradename
Rheodyne 6000-210
fitting
ferrule for 1/16 in. tubing
General description
The stainless steel ferrule is used for connecting most HPLC columns to a 1/16 in. tubing system.
Other Notes
fits Rheodyne nuts, p/n 58256 and 58257
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 1