Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

50653-U

Supelco

BIOshell A160 Peptide PCS-C18, 2.7 µm HPLC Column

column L × I.D. 50 mm × 1.5 mm, pk of 1 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709
NACRES:
SB.52
Pricing and availability is not currently available.

CofA

current certificate can be downloaded

Quality Level

packaging

pk of 1 ea

extent of labeling

5.1% Carbon loading

parameter

600 bar max. pressure (8700 psi)

maximum temperature

60 °C (pH2/60; pH7/40)

column L × I.D.

50 mm × 1.5 mm

surface area

75 m2/g

matrix

superficially porous particle particle platform (Fused-Core®)

matrix active group

C18 bonding phase, dimethyloctadecylsilane and positively charged ligand

operating pH range

2-7

Looking for similar products? Visit Product Comparison Guide

General description

BIOshell A160 Peptide PCS-C18 columns are specifically engineered to provide efficient separation of peptides as well as small proteins. These columns contain advanced Fused-Core® particles with pores strategically sized to 160 Å to enable optimized peptide diffusion. This attribute makes these columns an excellent choice for peptide mapping. The unique PCS-C18 chemistry enables exceptional peak shape and enhanced loading capacity of basic peptides as compared to traditional C18 chemistries. In addition, the PCS-C18 facilitates the use of mobile phases with lower concentrations of mobile phase modifiers without any compromise to column performance. This column also provides orthogonal selectivity to ES-C18, ES-CN (cyano), and phenyl-hexyl chemistries.

Analysis Note

Target analytes include peptides, proteins (< 25 kDa MW), tryptic digests, and post-translationally modified (PTM) peptides and polypeptides.

Legal Information

Fused-Core is a registered trademark of Advanced Materials Technology, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service