Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47517-U

Supelco

Kerosene Reference Standard

50,000 μg/mL in hexane, analytical standard

Synonym(s):

Kerosene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

vapor density

4.5 (vs air)

vapor pressure

0.23 mmHg ( 20 °C)

autoignition temp.

442 °F

feature

standard type calibration

expl. lim.

5 %

packaging

ampule of 1 mL

concentration

50,000 μg/mL in hexane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

190-250 °C (lit.)

density

0.8 g/mL at 25 °C (lit.)

application(s)

environmental
petroleum

format

single component solution

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

target_organs

Central nervous system, Nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

-14.8 °F - closed cup

flash_point_c

-26.0 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gerardo Ramos et al.
Toxicology and applied pharmacology, 195(3), 331-338 (2004-03-17)
Applying military jet fuel (JP-8) or commercial jet fuel (Jet-A) to the skin of mice suppresses the immune response in a dose-dependent manner. The release of biological response modifiers, particularly prostaglandin E2 (PGE2), is a critical step in activating immune
Kent L Gee et al.
The Journal of the Acoustical Society of America, 123(6), 4082-4093 (2008-06-10)
To address the question of the role of nonlinear effects in the propagation of noise radiated by high-power jet aircraft, extensive measurements were made of the F-22A Raptor during static engine run-ups. Data were acquired at low-, intermediate-, and high-thrust
Susan P Proctor et al.
Neurotoxicology, 32(6), 799-808 (2011-08-10)
One of the most prevalent workplace chemical exposures historically and currently confronting the global military and civilian workforce is jet propellant (JP) fuel (e.g., JP4, JP5, JP8, jet A1), a complex mixture of numerous hydrocarbon compounds and additives. To date
Gerardo Ramos et al.
Toxicological sciences : an official journal of the Society of Toxicology, 100(2), 415-422 (2007-09-25)
Dermal exposure to military (JP-8) and/or commercial (Jet-A) jet fuel suppresses cell-mediated immune reactions. Immune regulatory cytokines and biological modifiers, including platelet activating factor (PAF), prostaglandin E(2), and interleukin-10, have been implicated in the pathway of events leading to immune
J Rossi et al.
Journal of toxicology and environmental health. Part A, 63(6), 397-428 (2001-08-03)
The U.S. Naval Service is anticipating transition from the nearly exclusive use of JP-5 jet fuel to predominant use of JP-8, consistent with the primary utilization by the U.S. Army, U.S. Air Force, and the militaries of most NATO countries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service