Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442732

Supelco

p-Isopropyltoluene

analytical standard

Synonym(s):

p-Cymene, 1-Isopropyl-4-methylbenzene, 4-Isopropyltoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CH(CH3)2
CAS Number:
Molecular Weight:
134.22
Beilstein/REAXYS Number:
1903377
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor density

4.62 (vs air)

vapor pressure

1.5 mmHg ( 20 °C)
3.7 mmHg ( 37.7 °C)

autoignition temp.

817 °F

expl. lim.

5.6 %

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.490 (lit.)

bp

176-178 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

CC(C)c1ccc(C)cc1

InChI

1S/C10H14/c1-8(2)10-6-4-9(3)5-7-10/h4-8H,1-3H3

Inchi Key

HFPZCAJZSCWRBC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 3 - Repr. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

125.6 °F - closed cup

flash_point_c

52 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vanessa Kar-Yan Lo et al.
Journal of the American Chemical Society, 134(18), 7588-7591 (2012-04-26)
Complex [(p-cymene)Ru(η(1)-O(2)CCF(3))(2)(OH(2))] mediated transformation of α-diazoacetamides ArCH(2)N(C(CH(3))(3))C(O)CHN(2) to result in carbene insertion into the primary C-H bond exclusively, with the γ-lactam products being isolated in up to 98% yield. This unexpected reaction is striking in view of the presence of
Natalia Busto et al.
Chemistry, an Asian journal, 7(4), 788-801 (2012-02-07)
The reactions of two diaminotriazine ligands 2,4-diamino-6-(2-pyridyl)-1,3,5-triazine (2-pydaT) and 6-phenyl-2,4-diamino-1,3,5-triazine (PhdaT) with ruthenium-arene precursors led to a new family of ruthenium(II) compounds that were spectroscopically characterized. Four of the complexes were cationic, with the general formula [(η(6)-arene)Ru(κ(2)-N,N-2-pydaT)Cl]X (X=BF(4), TsO; arene=p-cymene:
Lutz Ackermann et al.
Organic letters, 14(8), 2146-2149 (2012-04-13)
The ruthenium(II) carboxylate complex [Ru(O(2)CMes)(2)(p-cymene)] enabled efficient direct arylations of unactivated C-H bonds with easily available, inexpensive phenols. Extraordinary chemoselectivity of the well-defined ruthenium catalyst set the stage for challenging C-H/C-O bond functionalizations to occur under solvent-free conditions as well
Lydia E H Paul et al.
Inorganic chemistry, 51(2), 1057-1067 (2012-01-10)
The relative affinity of the cationic triangular metallaprism, [(pCH(3)C(6)H(4)Pr(i))(6)Ru(6)(tpt)(2)(dhbq)(3)](6+) ([1](6+)), for various amino acids, ascorbic acid, and glutathione (GSH) has been studied at 37 °C in aqueous solutions at pD 7, using NMR spectroscopy and electrospray ionization mass spectrometry (ESI-MS).
Kenichi Ogata et al.
Dalton transactions (Cambridge, England : 2003), 42(7), 2362-2365 (2012-12-14)
A series of 1,2,3-triazol-5-ylidene (tzNHC) complexes of a (p-cymene)ruthenium system was synthesized. The reactivity of the N-bonded and C-bonded aryl groups in the tzNHC ligand was found to be significantly different with respect to intramolecular aromatic C-H bond activation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service