Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SPR-322B

StressMarq Biosciences Inc

StressMarq Alpha Synuclein Pre-formed Fibrils (Type 1)

100 µg

Synonym(s):

Alpha synuclein PFFs, Alpha synuclein aggregates, Non-A beta component of AD amyloid protein, NACP protein, SNCA protein, PARK1 protein, SYN protein, Parkinson disease familial 1 Protein

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$382.00

$382.00


Available to ship onApril 29, 2025Details



Select a Size

Change View
1 EA
$382.00

About This Item

Molecular Weight:
14.46
PDF Icon
Shipped & Sold By StressMarq Biosciences Inc.
A Trusted Marketplace Seller
PDF Icon
Have a question?

$382.00


Available to ship onApril 29, 2025Details


assay

95

concentration

2 mg/mL

application(s)

life science and biopharma

biological source

human

recombinant

E. coli

mol wt

predicted mol wt 14.46 kDa

Storage temp.

-80 °C

storage condition

avoid repeated freeze/thaw cycles

manufacturer/tradename

StressMarq Biosciences Inc

Related Categories

General description

Alpha Synuclein Pre-formed Fibrils (Type 1) are expressed in E. coli and Ion-exchange Purified. Certified >95% pure using SDS-PAGE analysis. Low endotoxin <5 EU/mL @ 2mg/mL. Validated for WB | SDS-PAGE | In vivo assay | In vitro assay. Monomer source is catalog# SPR-321.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library