Z653527
Pyrex® glass test tubes without rim, medium wall
O.D. × L 16 mm × 125 mm, wall thickness 1.2 mm
Synonym(s):
1622/10M
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass tube
colorless tube
straight neck
sterility
non-sterile
feature
graduations: no
Push fit
packaging
pack of 100 ea
manufacturer/tradename
SciLabware 1622/10M
availability
available only in EU
O.D. × L
16 mm × 125 mm
tube diam.
16 mm
wall thickness
1.2 mm
Looking for similar products? Visit Product Comparison Guide
General description
- Rimless for greater convenience in plugging and rack storage
- Ideal for culture work
- Fully autoclavable
- High resistance to attack by water
- Reduces leaching of contaminants which can cause pH changes
- Manufactured from heat resistant Pyrex borosilicate glass allowing use at high temperatures
- Conforms to ISO 4142
Legal Information
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service