Documents
Z601152
Hettich® centrifuge adapters
for 9-10 mL blood/urine tubes (15-16x92-102 mm) and 15 mL round bottom tubes (17x100 mm)
About This Item
UNSPSC Code:
41103911
NACRES:
NB.22
description
for EBA 21 and MIKRO 220/220R
feature
Holds 15 mL tubes
packaging
pack of 6 ea
manufacturer/tradename
Hettich 1635
technique(s)
centrifugation: suitable
compatibility
for use with 1016, 1137 (rotors)
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG