Key Documents
Z360961
Sigma-Aldrich
Sigma adjustable volume digital pipettes
capacity 50-200 μL
About This Item
UNSPSC Code:
41121500
capacity
50-200 μL
General description
Rugged, hand-held pipettes are excellent for student use, yet precise enough for demanding research applications. Comfortable one-hand operation with smooth plunger action and convenient tip ejector. Volume is continuously adjustable by rotating plunger button.
Each pipette has been tested and calibrated and comes with its own quality-control certificate.
comparable product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)