Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

WH0009842M1

Sigma-Aldrich

Monoclonal Anti-PLEKHM1 antibody produced in mouse

clone 1C9, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-AP162, Anti-B2, Anti-KIAA0356, Anti-pleckstrin homology domain containing, family M (with RUN domain) member 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$406.00

$406.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$406.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$406.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1C9, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable

isotype

IgG2bκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PLEKHM1(9842)

General description

The protein encoded by this gene is essential for bone resorption, and may play a critical role in vesicular transport in the osteoclast. Mutations in this gene are associated with autosomal recessive osteopetrosis type 6 (OPTB6). Alternatively spliced transcript variants have been found for this gene. (provided by RefSeq)

Immunogen

PLEKHM1 (AAH64361, 957 a.a. ~ 1056 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
PHRFSVADLQQIADGVYEGFLKALIEFASQHVYHCDLCTQRGFICQICQHHDIIFPFEFDTTVRCAECKTVFHQSCQAVVKKGCPRCARRRKYQEQNIFA

Biochem/physiol Actions

PLEKHM1 (pleckstrin homology and RUN domain containing M1) is involved in the end stages of endocytic and autophagy pathways. Here, it interacts with RAB7 (member of RAS oncogene family), the HOPS (homotypic fusion and vacuole protein sorting) complex and ATG8 (autophagy-related protein 8) proteins. It plays an important role in autophagosome-lysosome fusion and endosome and autophagosome maturation. PLEKHM1 also participates in osteoclast function and bone resorption. In presence of Salmonella enterica Typhimurium, the PLEKHM1-RAB7-HOPS complex targets the Salmonella effector protein SifA (Salmonella-induced filament protein A) and controls vacuoles consisting of Salmonella.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David G McEwan et al.
Autophagy, 11(4), 720-722 (2015-04-24)
The endosomal system and autophagy are 2 intertwined pathways that share a number of common protein factors as well as a final destination, the lysosome. Identification of adaptor platforms that can link both pathways are of particular importance, as they

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service