Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V4380

Sigma-Aldrich

[D-p-Cl-Phe6, Leu17]-Vasoactive Intestinal Peptide human, porcine, rat

≥97% (HPLC)

Synonym(s):

[D-p-Cl-Phe6, Leu17]-VIP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C148H239ClN44O42
CAS Number:
Molecular Weight:
3342.20
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

Quality Level

assay

≥97% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CCC(C)C(NC(=O)C(CO)NC(=O)C(CC(N)=O)NC(=O)C(CC(C)C)NC(=O)C(Cc1ccc(O)cc1)NC(=O)C(CCCCN)NC(=O)C(CCCCN)NC(=O)C(NC(=O)C(C)NC(=O)C(CC(C)C)NC(=O)C(CCC(N)=O)NC(=O)C(CCCCN)NC(=O)C(CCCNC(N)=N)NC(=O)C(CC(C)C)NC(=O)C(CCCNC(N)=N)NC(=O)C(NC(=O)C(Cc2ccc(O)cc2)NC(=O)C(CC(N)=O)NC(=O)C(CC(O)=O)NC(=O)C(NC(=O)C(Cc3ccc(Cl)cc3)NC(=O)C(NC(=O)C(C)NC(=O)C(CC(O)=O)NC(=O)C(CO)NC(=O)C(N)Cc4cnc[nH]4)C(C)C)C(C)O)C(C)O)C(C)C)C(=O)NC(CC(C)C)C(=O)NC(CC(N)=O)C(N)=O

InChI

1S/C148H239ClN44O42/c1-19-75(14)116(144(233)184-98(55-72(8)9)132(221)175-94(119(158)208)60-109(155)201)191-141(230)107(67-195)188-136(225)103(62-111(157)203)181-133(222)97(54-71(6)7)178-134(223)99(57-81-35-41-85(198)42-36-81)179-126(215)89(29-21-24-48-151)170-124(213)90(30-22-25-49-152)173-142(231)114(73(10)11)189-120(209)76(15)167-129(218)95(52-69(2)3)176-128(217)93(45-46-108(154)200)172-123(212)88(28-20-23-47-150)169-125(214)91(31-26-50-164-147(159)160)171-131(220)96(53-70(4)5)177-127(216)92(32-27-51-165-148(161)162)174-145(234)117(78(17)196)192-138(227)100(58-82-37-43-86(199)44-38-82)180-135(224)102(61-110(156)202)182-137(226)105(64-113(206)207)186-146(235)118(79(18)197)193-139(228)101(56-80-33-39-83(149)40-34-80)185-143(232)115(74(12)13)190-121(210)77(16)168-130(219)104(63-112(204)205)183-140(229)106(66-194)187-122(211)87(153)59-84-65-163-68-166-84/h33-44,65,68-79,87-107,114-118,194-199H,19-32,45-64,66-67,150-153H2,1-18H3,(H2,154,200)(H2,155,201)(H2,156,202)(H2,157,203)(H2,158,208)(H,163,166)(H,167,218)(H,168,219)(H,169,214)(H,170,213)(H,171,220)(H,172,212)(H,173,231)(H,174,234)(H,175,221)(H,176,217)(H,177,216)(H,178,223)(H,179,215)(H,180,224)(H,181,222)(H,182,226)(H,183,229)(H,184,233)(H,185,232)(H,186,235)(H,187,211)(H,188,225)(H,189,209)(H,190,210)(H,191,230)(H,192,227)(H,193,228)(H,204,205)(H,206,207)(H4,159,160,164)(H4,161,162,165)

InChI key

BUTRVBZATBJGPP-UHFFFAOYSA-N

Gene Information

human ... VIP(7432)

Amino Acid Sequence

His-Ser-Asp-Ala-Val-Cl-Phe-Thr-Asp-Asn-Tyr-Thr-Arg-Leu-Arg-Lys-Gln-Leu-Ala-Val-Lys-Lys-Tyr-Leu-Asn-Ser-Ile-Leu-Asn-NH2

Biochem/physiol Actions

VIP receptor antagonist

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Rekik et al.
The Journal of pharmacology and experimental therapeutics, 287(3), 832-838 (1998-12-24)
Distension of the small intestine can play a role in the pathogenesis of various functional intestinal disorders. This study determined the role of vasoactive intestinal polypeptide (VIP) in the adaptative response of intestinal smooth muscle to acute and chronic distension
Interaction of a sulfhydryl analogue of vasoactive intestinal peptide (VIP) with murine lymphocytes.
C A Ottaway
Advances in experimental medicine and biology, 371A, 553-558 (1995-01-01)
N P Xenopoulos et al.
The American journal of physiology, 266(2 Pt 2), H399-H405 (1994-02-01)
It is now recognized that stimulation of the vagus releases both acetylcholine (ACh) and vasoactive intestinal peptide (VIP). Whereas ACh depresses cardiac function, recent data indicate that VIP may have a cardiostimulatory effect. Exogenously administered VIP appears to enhance left
A Shvilkin et al.
Cardiovascular research, 28(12), 1769-1773 (1994-12-01)
The aim was to determine the extent to which endogenous release of vasoactive intestinal polypeptide (VIP) might be implicated in the modulation of sinoatrial rate in the presence and absence of muscarinic blockade or beta blockade. Langendorff perfused rat hearts
F Chang et al.
Circulation research, 74(1), 157-162 (1994-01-01)
We have investigated the actions of vasoactive intestinal peptide (VIP) and neuropeptide Y (NPY) on the pacemaker current (I(f)) in canine Purkinje fibers. On voltage pulses to the middle of the I(f) activation range, VIP reversibly increases I(f), whereas NPY

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service