Key Documents
TT38440SB-EP
Eppendorf twin.tec® PCR Plates 384
skirted, PCR clean, volume 40 μL, colorless wells, blue plate, pack of 25
Select a Size
25 EA
$292.00
$292.00
Available to ship onApril 22, 2025Details
Select a Size
Change View
25 EA
$292.00
About This Item
UNSPSC Code:
41106300
material:
blue plate
colorless wells
polypropylene wells
size:
384 well
sterility:
non-sterile
feature:
skirt
PCR clean
skirted
$292.00
Available to ship onApril 22, 2025Details
material
blue plate
colorless wells
polypropylene wells
well size
384
sterility
non-sterile
feature
skirt
PCR clean
skirted
packaging
pack of 25
manufacturer/tradename
Eppendorf® 951020737 (NA)
technique(s)
PCR: suitable
size
384 well
volume
40 μL
working volume
40 μL
General description
The Eppendorf twin.tec PCR plate combines extremely thin-walled polypropylene wells for optimal heat transfer to the sample, with robust polycarbonate frames for high rigidity. With raised well rims, twin.tec plates enable highly effective sealing to reduce the risk of cross-contamination.
Features and Benefits
  • Optimized rim for barcodes and automated work processes
  • StackableWithin SBS footprint recommendationsI
  • deal for most common 384-well thermocyclers, particularly for Eppendorf Mastercycler X50 and Mastercycler nexus
  • 40 µL max. well volume
  • Ideal for quantitative real-time PCR as well – OptiTrack matrix for faster sample identification and fewer pipetting errors
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)