Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T7068

Sigma-Aldrich

Thymeleatoxin

99%, solid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H36O10
CAS Number:
Molecular Weight:
628.67
MDL number:
UNSPSC Code:
12352200

assay

99%

form

solid

color

white

solubility

DMSO: soluble
H2O: insoluble
ethanol: soluble

storage temp.

−20°C

InChI

1S/C36H36O10/c1-19(2)34-28(42-25(38)16-15-22-11-7-5-8-12-22)21(4)35-24-17-20(3)27(39)33(24,41)31(40)32(18-37)29(43-32)26(35)30(34)44-36(45-34,46-35)23-13-9-6-10-14-23/h5-17,21,24,26,28-31,37,40-41H,1,18H2,2-4H3/b16-15+/t21-,24-,26+,28-,29+,30-,31-,32+,33-,34+,35-,36?/m1/s1

InChI key

OTTFLYUONKAFGT-WOYWLKGSSA-N

Biochem/physiol Actions

Selective activator of PKC isotypes α, β1 and γ

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inês E Baldeiras et al.
Biological research, 39(3), 531-539 (2006-11-16)
Thymeleatoxin (TMX), an activator of Ca2+-sensitive protein kinase C (cPKC) isoforms, was used to assess the PKC isoform specificity of cholinergic potentiation of glucose (11 mM)-induced pulsatile 5-HT/insulin release (PIR) from single mouse pancreatic islets. TMX (100 nM) and carbachol
Bokyung Kim et al.
British journal of pharmacology, 139(2), 408-414 (2003-05-29)
1 Phorbol ester decreases muscle tension in the rat myometrium, and the effect is more potent in late-pregnant myometrium than in nonpregnant myometrium. In the present study, we have examined the contribution of protein kinase C (PKC) isoforms to the
Ann-Kathrin Rahm et al.
British journal of pharmacology, 166(2), 764-773 (2011-12-16)
Two-pore-domain K(+) channels (K(2P) ) mediate K(+) background currents that modulate the membrane potential of excitable cells. K(2P) 18.1 (TWIK-related spinal cord K(+) channel) provides hyperpolarizing background currents in neurons. Recently, a dominant-negative loss-of-function mutation in K(2P) 18.1 has been
Dierk Thomas et al.
Cardiovascular research, 59(1), 14-26 (2003-06-28)
Patients with HERG-associated long QT syndrome typically develop tachyarrhythmias during physical or emotional stress. Previous studies have revealed that activation of the beta-adrenergic system and consecutive elevation of the intracellular cAMP concentration regulate HERG channels via protein kinase A-mediated phosphorylation
Youn Ri Lee et al.
Journal of pharmacological sciences, 100(2), 119-125 (2006-02-14)
Vascular smooth muscle contraction is mediated by activation of extracellular signal-regulated kinase (ERK) 1/2, an isoform of mitogen-activated protein kinase (MAPK). However, the role of stress-activated protein kinase/c-Jun N-terminal kinase (JNK) in vascular smooth muscle contraction has not been defined.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service