Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T6303

Sigma-Aldrich

Bis(trimethylsilyl) phosphite

≥98% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H19O3PSi2
CAS Number:
Molecular Weight:
226.36
Beilstein/REAXYS Number:
1776928
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥98% (GC)

density

0.968 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

C[Si](C)(C)OP(O)O[Si](C)(C)C

InChI

1S/C6H19O3PSi2/c1-11(2,3)8-10(7)9-12(4,5)6/h7H,1-6H3

InChI key

TVYLOQJEHUAACN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service