Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T5271

Sigma-Aldrich

1,1,2-Trichloro-1,2,2-trifluoroethane

Synonym(s):

1,1,2-Trichloro-trifluoroethane, Genetron® 113

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCF2CCl2F
CAS Number:
Molecular Weight:
187.38
Beilstein/REAXYS Number:
1740335
EC Number:
MDL number:
UNSPSC Code:
12352200

vapor pressure

5.5 psi ( 20 °C)

autoignition temp.

1256 °F

refractive index

n20/D 1.358 (lit.)

bp

47-48 °C (lit.)

mp

−35 °C (lit.)

density

1.57 g/mL at 25 °C (lit.)
1.58 g/mL at 25 °C (lit.)

SMILES string

FC(F)(Cl)C(F)(Cl)Cl

InChI

1S/C2Cl3F3/c3-1(4,6)2(5,7)8

InChI key

AJDIZQLSFPQPEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Genetron is a registered trademark of Honeywell International Inc.

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Ozone 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

383.0 °F

flash_point_c

195 °C

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service