Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T1515

Sigma-Aldrich

Tetraethylammonium acetate tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)4N(OCOCH3) · 4H2O
CAS Number:
Molecular Weight:
261.36
Beilstein/REAXYS Number:
3574285
EC Number:
MDL number:
UNSPSC Code:
12352200

mp

42-46 °C (lit.)

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].CC([O-])=O.CC[N+](CC)(CC)CC

InChI

1S/C8H20N.C2H4O2.4H2O/c1-5-9(6-2,7-3)8-4;1-2(3)4;;;;/h5-8H2,1-4H3;1H3,(H,3,4);4*1H2/q+1;;;;;/p-1

InChI key

QNOAKQGNSWGYOE-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3

flash_point_f

140.0 °F - closed cup

flash_point_c

60 °C - closed cup

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service