Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T105

Sigma-Aldrich

Tetraethylammonium chloride

Synonym(s):

TEA chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)4N(Cl)
CAS Number:
Molecular Weight:
165.70
Beilstein/REAXYS Number:
3563247
EC Number:
MDL number:
UNSPSC Code:
12161900

SMILES string

[Cl-].CC[N+](CC)(CC)CC

InChI

1S/C8H20N.ClH/c1-5-9(6-2,7-3)8-4;/h5-8H2,1-4H3;1H/q+1;/p-1

InChI key

YMBCJWGVCUEGHA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

201.2 °F - closed cup

flash_point_c

94 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takahiro Shibayama et al.
Pharmaceutical research, 32(9), 2937-2949 (2015-03-21)
Unstirred water layers (UWLs) present an unavoidable complication to the measurement of transport kinetics in cultured cells, and the high rates of transport achieved by overexpressing heterologous transporters exacerbate the UWL effect. This study examined the correlation between measured Jmax
Fernanda S Gravina et al.
PloS one, 9(10), e111307-e111307 (2014-10-23)
The pacemaker mechanisms activating phasic contractions of vaginal and cervical smooth muscle remain poorly understood. Here, we investigate properties of pacemaking in vaginal and cervical tissues by determining whether: 1) functional pacemaking is dependent on the phase of the estrus
Nanomechanics of cation-π interactions in aqueous solution.
Qingye Lu et al.
Angewandte Chemie (International ed. in English), 52(14), 3944-3948 (2013-03-02)
Min Shen et al.
Vascular pharmacology, 58(1-2), 64-70 (2012-07-24)
Evidence has indicated that resveratrol (Res) produces vasorelaxation and may decrease the coronary heart disease mortality. However, several pathways involved in the mechanism of vasorelaxation are still unclear. This study was designed, therefore, to test the probable ion channels or
Karen Mruk et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(34), 13579-13583 (2012-08-08)
Calmodulin (CaM) is a ubiquitous intracellular calcium sensor that directly binds to and modulates a wide variety of ion channels. Despite the large repository of high-resolution structures of CaM bound to peptide fragments derived from ion channels, there is no

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service