About This Item
assay
≥98%
mp
161-165 °C (lit.)
SMILES string
Cl[H].N[C@H](CO)Cc1ccc(O)cc1
InChI
1S/C9H13NO2.ClH/c10-8(6-11)5-7-1-3-9(12)4-2-7;/h1-4,8,11-12H,5-6,10H2;1H/t8-;/m0./s1
InChI key
PNGCRFWYSRUQTB-QRPNPIFTSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service