Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T0389

Sigma-Aldrich

2,2,2-Trichloroethanol

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352100

density

1.53 g/mL at 25 °C (lit.)

replaced by

Product No.
Description
Pricing

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service