Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6163

Sigma-Aldrich

FLT-3 Ligand human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

CD135, FLK2, Fms-related tyrosine kinase 3 ligand, STK1, Stem cell tyrosine kinase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

24-30 kDa (monomer, glycosylated)

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... FLT3(2322)

General description

FLT3 ligand is a receptor for the Flt3 cytokine and has a tyrosine-protein kinase activity & a growth factor that regulates proliferation of early hematopoietic cells. Flt3-Ligand synergizes with other CSFs and interleukins to induce growth and differentiation. Human flt3-Ligand (flk2-ligand) stimulates the proliferation and colony formation of certain bone marrow precursor cells including CD34+ cells.

Physical form

Lyophilized from a PBS solution.

Preparation Note

Centrifuge the vial prior to opening.

Reconstitution

Reconstitute in sterile PBS containing 0.1% endotoxin-free, recombinant human serum albumin.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service