Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3793

Sigma-Aldrich

MS4322

≥95% (HPLC)

Synonym(s):

N-((S)-3-(3,4-Dihydroisoquinolin-2(1H)-yl)-2-hydroxypropyl)-6-((1-((S)-19-((2S,4R)-4-hydroxy-2-(((S)-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl)carbamoyl)pyrrolidine-1-carbonyl)-20,20-dimethyl-17-oxo-3,6,9,12,15-pentaoxa-18-azahenicosanoyl)azetidin-3-yl)amino)pyrimidine-4-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C55H76N10O12S
CAS Number:
Molecular Weight:
1101.32
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Potent, cell-permeable, highly selective and yet a slow PRMT5 degrader.

MS4322 is a potent, cell-permeable, highly selective and yet a slow PRMT5 (protein arginine methyltransferase 5) degrader. It comprises of PRMT5 inhibitor EPZ015666 linked to a von Hippel-Lindau (VHL) E3 ligase ligand (S,R,S)-AHPC-Me (VHL-2). MS4322 reduces PRMT5 protein levels in a concentration-, time-, VHL- and proteasome-dependent manner. Inhibits PRMT5 activity (IC50 = 18 nM) and reduces arginine symmetric dimethylation (SDMA) levels. MS4322 displays good plasma exposure in a mouse PK study and in vivo availability.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yudao Shen et al.
Journal of medicinal chemistry, 63(17), 9977-9989 (2020-08-14)
The aberrant expression of protein arginine methyltransferase 5 (PRMT5) has been associated with multiple cancers. Using the proteolysis targeting chimera technology, we discovered a first-in-class PRMT5 degrader 15 (MS4322). Here, we report the design, synthesis, and characterization of compound 15

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service