Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SML3769

Sigma-Aldrich

CWI1-2 Hydrochloride

≥98% (HPLC)

Synonym(s):

N,N′,N′′-Tris(5-chloro-2-hydroxybenzylideneamino)guanidine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H17Cl3N6O3·HCl
CAS Number:
Molecular Weight:
556.23
UNSPSC Code:
12352200
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC1=CC=C(O)C(/C=N/N=C(N/N=C/C2=C(O)C=CC(Cl)=C2)N/N=C/C3=C(O)C=CC(Cl)=C3)=C1.Cl

InChI key

UXAOLDYLYMMMRC-SXOKSYETSA-N

Biochem/physiol Actions

CWI1-2 is a cell penetrant, potent and selective inhibitor of an m6A reader IGF2BP2 (Insulin-like growth factor 2 mRNA-binding protein 2). CWI1-2 potently inhibits IGF2BP2 interaction with N6-Methyladenosine (m6A) modified target transcripts. CWI1-2 exhibits potent anti-leukemia efficacy in vitro and in vivo.
Cell penetrant, potent and selective inhibitor of an m6A reader IGF2BP2

Caution

Hygroscopic

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Novel N,N?,N?-Tris[aryl(hetaryl)-methylideneamino]guanidine Derivatives as Efficient and Selective Colorimetric Sensors for Fluoride Ion
Russian Journal of Organic chemistry (2019)
The m6A reader IGF2BP2 regulates glutamine metabolism and represents a therapeutic target in acute myeloid leukemia
Cancer Cell, 40(12), 1566-1582 (2022)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service