Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SML3748

Sigma-Aldrich

TH10785

≥98% (HPLC)

Synonym(s):

N-Cyclohexyl-2-cyclopropyl-4-quinazolinamine, NN-Cyclohexyl-2-cyclopropylquinazolin-4-amine, TH 10785, TH-10785, V028-5832

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21N3
CAS Number:
Molecular Weight:
267.37
UNSPSC Code:
12352200
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

C1(N=C(C2CC2)N=C3NC4CCCCC4)=C3C=CC=C1

InChI key

ZQMGQZOHIDOPCQ-UHFFFAOYSA-N

Biochem/physiol Actions

8-oxoG DNA glycosylase OGG1 activator that induced β,δ-lyase activity for polynucleotide kinase phosphatase (PNKP1)-dependent base excision repair (BER).
TH10785 is an 8-oxoguanine (8-oxoG) DNA glycosylase 1 (OGG1) activator that interats with OGG1 via Phe319 and Gly42, causing increased OGG1 recruitment to cellular oxidative DNA damage sites (by 1.37-fold post 20 sec 1 µM treatment; U2OS). TH10785-activated OGG1 recognizes not only 8-oxoG, but also apurinic/apyrimidinic (AP, abasic) sites with a higher efficiency due to a newly induced β,δ-lyase enzymatic activity that generates base excision repair (BER) intermediates that require polynucleotide kinase phosphatase (PNKP1) instead of apurinic endonuclease 1 (APE1) for completing the repair.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beverly A Baptiste et al.
Free radical biology & medicine, 124, 149-162 (2018-06-08)
The common oxidatively generated lesion, 8-oxo-7,8-dihydroguanine (8-oxoGua), is removed from DNA by base excision repair. The glycosylase primarily charged with recognition and removal of this lesion is 8-oxoGuaDNA glycosylase 1 (OGG1). When left unrepaired, 8-oxodG alters transcription and is mutagenic.
A "Failed" Assay Development for the Discovery of Rescuing Small Molecules from the Radiation Damage
SLAS discovery, 26(10), 1315-1325 (2021)
Maurice Michel et al.
Science (New York, N.Y.), 376(6600), 1471-1476 (2022-06-24)
Oxidative DNA damage is recognized by 8-oxoguanine (8-oxoG) DNA glycosylase 1 (OGG1), which excises 8-oxoG, leaving a substrate for apurinic endonuclease 1 (APE1) and initiating repair. Here, we describe a small molecule (TH10785) that interacts with the phenylalanine-319 and glycine-42

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service