Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3535

Sigma-Aldrich

GI-530159

≥98% (HPLC)

Synonym(s):

2,2-Bis[4-(4-aminophenoxy)phenyl]-1,1,1,3,3,3-hexafluoropropane, 2,2-Bis[4-(4-aminophenoxy)phenyl]hexafluoropropane, 4,4′-(Hexafluoroisopropylidene)bis(p-phenyleneoxy)dianiline, 4,4′-[[2,2,2-Trifluoro-1-(trifluoromethyl)ethylidene]bis(4,1-phenyleneoxy)]bis[benzenamine]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H20F6N2O2
CAS Number:
Molecular Weight:
518.45
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(F)(F)C(C(F)(F)F)(c3ccc(cc3)Oc4ccc(cc4)N)c1ccc(cc1)Oc2ccc(cc2)N

InChI

1S/C27H20F6N2O2/c28-26(29,30)25(27(31,32)33,17-1-9-21(10-2-17)36-23-13-5-19(34)6-14-23)18-3-11-22(12-4-18)37-24-15-7-20(35)8-16-24/h1-16H,34-35H2

InChI key

HHLMWQDRYZAENA-UHFFFAOYSA-N

Biochem/physiol Actions

Potent and selective opener of TWIK-related K (TREK) channels

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

GI-530159, a novel, selective, mechanosensitive two-pore-domain potassium (K2P ) channel opener, reduces rat dorsal root ganglion neuron excitability
British Journal of Pharmacology, 175(12), 2272-2283 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service