Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3377

Sigma-Aldrich

BG-3

≥98% (HPLC)

Synonym(s):

N-(4-Chlorophenyl)-6-ethyl-5-methyl[1,2,4]triazolo[1,5-a]pyrimidin-7-amine, SID 124738112

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14ClN5
CAS Number:
Molecular Weight:
287.75
UNSPSC Code:
51111800
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=NC2=NC=NN2C(NC3=CC=C(Cl)C=C3)=C1CC

Biochem/physiol Actions

BG-3 is a cell penetrant, potent and selective inhibitor of Pax2 (paired-box 2) that inhibits Pax2 transactivation ability. BG-3 inhibit the ability of Pax proteins to recruit PTIP and promote H3K4 methylation at Pax binding sites. BG-3 is not cytotoxic.
cell penetrant, potent and selective inhibitor of Pax2 that inhibits Pax2 transactivation ability

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shayna T J Bradford et al.
Cell chemical biology, 29(3), 412-422 (2021-11-26)
The Pax family of developmental control genes are frequently deregulated in human disease. In the kidney, Pax2 is expressed in developing nephrons but not in adult proximal and distal tubules, whereas polycystic kidney epithelia or renal cell carcinoma continues to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service