Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3027

Sigma-Aldrich

FMF-04-159-2

≥98% (HPLC)

Synonym(s):

(E)-N-(1-((3-(4-(dimethylamino)but-2-enamido)phenyl)sulfonyl)piperidin-4-yl)-4-(2,4,6-trichlorobenzamido)-1H-pyrazole-3-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$95.95
25 MG
$407.00

$95.95

List price$101.00
Web-Only Promotion

Available to ship onApril 08, 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
$95.95
25 MG
$407.00

About This Item

Empirical Formula (Hill Notation):
C28H30Cl3N7O5S
CAS Number:
Molecular Weight:
683.01
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$95.95

List price$101.00
Web-Only Promotion

Available to ship onApril 08, 2025Details


Request a Bulk Order

Quality Level

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=NNC=C1NC(C2=C(Cl)C=C(Cl)C=C2Cl)=O)NC3CCN(S(=O)(C4=CC=CC(NC(/C=C/CN(C)C)=O)=C4)=O)CC3

InChI key

HZPYSAHDRSBARR-QPJJXVBHSA-N

Biochem/physiol Actions

FMF-04-159-2 is a specific and potent covalent inhibitor Cyclin-dependent kinase 14 (CDK14) in cells that targets CDK14 at C218. FMF-04-159-2 reversibly inhibits CDK2, CDK10, and other TAIRE kinases (CDK15, CDK16, CDK17, and CDK18) with comparable potency to each other. It exhibits mild antiproliferative effects.
specific and potent covalent inhibitor CDK14 in cells that targets CDK14 at C218

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service