Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1493

Sigma-Aldrich

Suprafenacine

≥98% (HPLC)

Synonym(s):

4,5,6,7-Tetrahydro-1H-indazole-3-carboxylic acid (2E)-2-[(4-methylphenyl)methylene]hydrazide, 4,5,6,7-Tetrahydro-1H-indazole-3-carboxylic acid [1p-tolyl-meth-(E)-ylidene]-hydrazide, SRF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18N4O
CAS Number:
Molecular Weight:
282.34
UNSPSC Code:
12352202
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C16H18N4O/c1-11-6-8-12(9-7-11)10-17-20-16(21)15-13-4-2-3-5-14(13)18-19-15/h6-10H,2-5H2,1H3,(H,18,19)(H,20,21)/b17-10+

InChI key

WKKLZIQHOXPLLZ-LICLKQGHSA-N

Biochem/physiol Actions

Suprafenacine is a cell permeable and potent microtubule destabilizing agent that induces cell cycle arrest in the G2/M phase and apoptotic cell death. Suprafenacine binds to microtubules at the colchicine-binding and inhibits polymerization. Suprafenacine is selective for cancer cells including drug-resistant cancer cells. Apparently, Suprafenacine bypasses the multidrug resistance transporter P-glycoprotein (PGP).
Suprafenacine is a cell permeable and potent microtubule destabilizing agent.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service