Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1197

Sigma-Aldrich

ML 00253764 hydrochloride

≥98% (HPLC)

Synonym(s):

2-[2-[2-(5-Bromo-2- methoxyphenyl)-ethyl]-3-fluorophenyl]-4,5-dihydro-1H-imidazole hydrochloride, ML-00253764 hydrochloride, ML00253764 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18BrFN2O · HCl
CAS Number:
Molecular Weight:
413.71
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

FC1=C(CCC2=C(OC)C=CC(Br)=C2)C(C3=NCCN3)=CC=C1.[H]Cl

InChI

1S/C18H18BrFN2O.ClH/c1-23-17-8-6-13(19)11-12(17)5-7-14-15(3-2-4-16(14)20)18-21-9-10-22-18;/h2-4,6,8,11H,5,7,9-10H2,1H3,(H,21,22);1H

InChI key

KZUMGPQDDCBFBF-UHFFFAOYSA-N

Biochem/physiol Actions

ML 00253764 is a brain penetrant and potent MC4 melanocortin receptor antagonist that increases food intake and reduces loss of lean body mass in tumor bearing mice.
ML 00253764 is a brain penetrant and potent MC4 melanocortin receptor antagonist.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service