Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1021

Sigma-Aldrich

BC-1258

Synonym(s):

N,N′-Bis-(4-thiazol-2-yl-benzyl)-ethane-1,2-diamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H22N4S2
CAS Number:
Molecular Weight:
406.57
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

C1(C2=NC=CS2)=CC=C(CNCCNCC3=CC=C(C4=NC=CS4)C=C3)C=C1

InChI

1S/C22H22N4S2/c1-5-19(21-25-11-13-27-21)6-2-17(1)15-23-9-10-24-16-18-3-7-20(8-4-18)22-26-12-14-28-22/h1-8,11-14,23-24H,9-10,15-16H2

InChI key

FRTSLZJCGAFSET-UHFFFAOYSA-N

Biochem/physiol Actions

BC-1258 is a potent activator of unique E3 ligase F-box/LRR-repeat protein 2 (FBXL2) that increases levels of FBXLs protein, thus promoting Aurora B degradation. BC-1258 induces mitotic arrest and apoptosis of tumorigenic cells. Also BC-1258 inhibits tumor formation in athymic nude mice.
BC-1258 is a potent activator of unique E3 ligase F-box/LRR-repeat protein 2 (FBXL2).

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service