Skip to Content
MilliporeSigma

SML1007

B355252

≥98% (HPLC)

Synonym(s):

4-Chloro-N-(naphthalen-1-ylmethyl)-5-(3-(piperazin-1-yl)phenoxy)thiophene-2-sulfonamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H24ClN3O3S2
CAS Number:
Molecular Weight:
514.06
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

Biochem/physiol Actions

B355252 is a neuroprotective agent that potentiates nerve growth factor (NGF)-induced neurite outgrowth and protects against cell death caused by glutamate-evoked oxidative stress.
B355252 is a neuroprotective agent that potentiates nerve growth factor (NGF)-induced neurite outgrowth and protects against cell death caused by glutamate-evoked oxidative stress. In a murine hippocampal neuronal cell line, B355252 increased cell viability and protected against cell death caused by glutamate-induced toxicity, inhibiting glutamate-evoked increase in intracellular Ca2+ and ROS production.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library