Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0980

Sigma-Aldrich

CYM51010

≥98% (HPLC)

Synonym(s):

1-[[4-(Acetylamino)phenyl]methyl]-4-(2-phenylethyl)-4-Piperidinecarboxylic acid ethyl ester, CID23723457, ML335

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$88.40
25 MG
$519.00

$88.40


Estimated to ship onMay 03, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$88.40
25 MG
$519.00

About This Item

Empirical Formula (Hill Notation):
C25H32N2O3
CAS Number:
Molecular Weight:
408.53
UNSPSC Code:
12352200
NACRES:
NA.77

$88.40


Estimated to ship onMay 03, 2025


Request a Bulk Order

Quality Level

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

CYM51010 is a biased ligand of μ-opioid receptor - δ-opioid receptor heterodimers.
CYM51010 is a biased ligand of μ-opioid receptor – δ-opioid receptor heterodimers. CYM51010 exhibits anti-nociceptive activity similar to morphine, but with a decreased levels of tolerance development and withdrawal symptoms.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service