Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML0962

Sigma-Aldrich

M-110

≥98% (HPLC)

Synonym(s):

2-[[3-(4-Morpholinyl)propyl]amino]-4-pyridinecarboxylic acid 2-[1-(4-chloro-2-hydroxyphenyl)propylidene]hydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28ClN5O3
CAS Number:
Molecular Weight:
445.94
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

M-110 is a highly isoform-selective, cell permeable and potent ATP-competitive inhibitor of the PIM kinase family that prefers PIM-3. M-110 is an inhibitor of Wnt/β-catenin signaling that act downstream of the symb-catenin destruction complex to inhibit both Wnt-induced and cancer associated constitutive Wnt signaling via destabilization of β-catenin. The compound chelates iron in vitro and in intact cells. M-110 inhibits the proliferation of prostate cancer cell lines with IC50s of 0.6 to 0.9 μM, with no activity on normal human peripheral blood mononuclear cells up to 40 μM.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service