Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0960

Sigma-Aldrich

KKL-35

≥98% (HPLC)

Synonym(s):

4-Chloro-N-[5-(4-fluorophenyl)-1,3,4-oxadiazol-2-yl]-benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H9ClFN3O2
CAS Number:
Molecular Weight:
317.70
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

KKL-35 is a small molecule that blocks bacterial trans-translation, a proof-reading mechanism that eliminates mRNAs lacking stop codons from translation complexes. KKL-35 inhibits trans-translation, and subsequent cell growth in a broad spectrum of bacterial strains.
KKL-35 is an inhibitor of bacterial trans-translation.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service