Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0823

Sigma-Aldrich

DIMN

≥98% (HPLC)

Synonym(s):

6-(3,4-Dihydro-1H-isoquinolin-2-yl)-N-(6-methylpyridin-2-yl)nicotinamide, 6-(3,4-Dihydro-2(1H)-isoquinolinyl)-N-(6-methyl-2-pyridinyl)-3-pyridinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20N4O
CAS Number:
Molecular Weight:
344.41
UNSPSC Code:
51111800
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

DIMN is an androgen receptor (AR) antagonist.
DIMN is an androgen receptor (AR) antagonist. DIMN is an anti-androgen that effectively suppresses growth of both androgen-dependent (LNCaP) and androgen-independent prostate cancer cell lines (C4-2 and CWR22rv) that express AR.

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service