Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0787

Sigma-Aldrich

AC 42

≥98% (HPLC)

Synonym(s):

4-(4-Butyl-1-piperidinyl)-1-(2-methylphenyl)-1-butanone hydrochloride, AC-42

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$103.00

$103.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$103.00

About This Item

Empirical Formula (Hill Notation):
C20H31NO · HCl
CAS Number:
Molecular Weight:
337.93
UNSPSC Code:
12352202
NACRES:
NA.77

$103.00


Check Cart for Availability

Request a Bulk Order

Quality Level

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

Cl.N2(CCC(CC2)CCCC)CCCC(=O)c1c(cccc1)C

InChI

1S/C20H31NO.ClH/c1-3-4-9-18-12-15-21(16-13-18)14-7-11-20(22)19-10-6-5-8-17(19)2;/h5-6,8,10,18H,3-4,7,9,11-16H2,1-2H3;1H

InChI key

HJCOVULGUXGGSU-UHFFFAOYSA-N

Biochem/physiol Actions

AC-42 is a specific agonist of the muscarininc M1 receptor (pEC50 = 6.54) with little or no activity against other muscarinic receptor family members. AC-42 binds to an allosteric site adjacent to the adjacent to the orthosteric, acetylcholine binding site.
AC-42 is a specific agonist of the muscarininc M1 receptor.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beatrice Mihaela Radu et al.
Scientific reports, 7(1), 5083-5083 (2017-07-13)
Clinical and experimental studies indicate that muscarinic acetylcholine receptors are potential pharmacological targets for the treatment of neurological diseases. Although these receptors have been described in human, bovine and rat cerebral microvascular tissue, a subtype functional characterization in mouse brain

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service