Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0427

Sigma-Aldrich

Cardionogen-2

≥98% (HPLC)

Synonym(s):

6-(3,4-Dimethoxyphenyl)-3-(2-pyridinyl)-1,2,4-triazolo[3,4-b][1,3,4]thiadiazole, 6-(3,4-Dimethoxyphenyl)-3-pyridin-2-yl-[1,2,4]triazolo[3,4-b][1,3,4]thiadiazole, CDNG2, vuc198

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13N5O2S
CAS Number:
Molecular Weight:
339.37
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL (clear solution; warmed)

storage temp.

2-8°C

SMILES string

COc1ccc(cc1OC)-c2nn3c(nnc3s2)-c4ccccn4

InChI

1S/C16H13N5O2S/c1-22-12-7-6-10(9-13(12)23-2)15-20-21-14(18-19-16(21)24-15)11-5-3-4-8-17-11/h3-9H,1-2H3

InChI key

MEWRLQUBVOVSGN-UHFFFAOYSA-N

Biochem/physiol Actions

Cardionogen-2 is a biphasic modulator of cardiogenesis that enlarges the size of the zebrafish embryonic heart by promoting cardiomyocyte formation. Also, cardionogen-2 promotes murine ES cells to differentiate into beating cardiomyocytes. Apparently, cardionogen-2 inhibits Wnt/b-catenin dependent transcriptional activity. Treatment of zebrafish embryos with cardionogen-2 significantly enlarged both the atrium and ventricle, without causing apparent defects of embryonic morphology or other organ development.
Cardionogen-2 is a biphasic modulator of cardiogenesis; inhibits Wnt/β-catenin dependent transcriptional activity.

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service