Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0373

Sigma-Aldrich

CZC-25146 hydrochloride

≥98% (HPLC)

Synonym(s):

N-[2-[[5-Fluoro-2-[[2-methoxy-4-(4-morpholinyl)phenyl]amino]-4-pyrimidinyl]amino]phenyl]-methanesulfonamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H25FN6O4S · HCl
CAS Number:
Molecular Weight:
525.00
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: >15 mg/mL (warmed)

storage temp.

2-8°C

SMILES string

Cl.COc1cc(ccc1Nc2ncc(F)c(Nc3ccccc3NS(C)(=O)=O)n2)N4CCOCC4

InChI

1S/C22H25FN6O4S.ClH/c1-32-20-13-15(29-9-11-33-12-10-29)7-8-19(20)26-22-24-14-16(23)21(27-22)25-17-5-3-4-6-18(17)28-34(2,30)31;/h3-8,13-14,28H,9-12H2,1-2H3,(H2,24,25,26,27);1H

InChI key

SKYQTYQDKAOHLP-UHFFFAOYSA-N

Biochem/physiol Actions

CZC-25146 is a potent selective LRRK2 inhibitor.
CZC-25146 is a potent selective and metabolically stable inhibitor of leucine-rich repeat kinase-2 (LRRK2) also known as dardarin or PARK8, a kinase with mutations linked to Parkinson′s disease. Because the most common mutation, G2019S, enhances LRRK2 kinase activity, it is hoped that LRRK2 inhibitors may be useful in treating the disease. CZC-25146 was shown to prevent mutant LRRK2-induced injury of cultured rodent and human neurons with mid-nanomolar potency.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service