Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0262

Sigma-Aldrich

GSK575594A

≥98% (HPLC)

Synonym(s):

1-(4-Amino-2-fluorophenyl)-4-[[4′-fluoro-4-(methylsulfonyl)[1,1′-biphenyl]-2-yl]carbonyl]-piperazine (9CI), [4-(4-Amino-2-fluorophenyl)-1-piperazinyl][4′-fluoro-4-(methylsulfonyl)[1,1′-biphenyl]-2-yl]-methanone

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$470.00

$470.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
25 MG
$470.00

About This Item

Empirical Formula (Hill Notation):
C24H23F2N3O3S
CAS Number:
Molecular Weight:
471.52
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$470.00


In StockDetails


Request a Bulk Order

assay

≥98% (HPLC)

form

powder

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

color

white to beige

solubility

DMSO: ≥5 mg/mL

storage temp.

2-8°C

SMILES string

CS(=O)(=O)c1ccc(-c2ccc(F)cc2)c(c1)C(=O)N3CCN(CC3)c4ccc(N)cc4F

InChI

1S/C24H23F2N3O3S/c1-33(31,32)19-7-8-20(16-2-4-17(25)5-3-16)21(15-19)24(30)29-12-10-28(11-13-29)23-9-6-18(27)14-22(23)26/h2-9,14-15H,10-13,27H2,1H3

InChI key

DAPKZHQMEQRNPY-UHFFFAOYSA-N

Biochem/physiol Actions

GSK575594A is a selective agonist of human GPR55.
GSK575594A is a selective agonist of human GPR55. It is specific for human over rodent GPR55. GSK575594A had no activity across a set of more than 200 validated molecular target assays from diverse classes including kinases, proteases and other enzymes, GPCRs, ion channels, nuclear receptors, and membrane transporters.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service