Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00544

Sigma-Aldrich

8H-1,3-Dioxolo[4,5-g][1]benzopyran-8-one, 7,9-dimethoxy-6-(4-methoxyphenyl)- (9CI, ACI)

≥95% (LC/MS-UV)

Synonym(s):

7,9-Dimethoxy-6-(4-methoxyphenyl)-8H-1,3-dioxolo[4,5-g][1]benzopyran-8-one (ACI), 8H-1,3-Dioxolo[4,5-g][1]benzopyran-8-one, 7,9-dimethoxy-6-(p-methoxyphenyl)- (7CI)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$444.00

$444.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
1 MG
$444.00

About This Item

Empirical Formula (Hill Notation):
C19H16O7
CAS Number:
Molecular Weight:
356.33
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

$444.00


Usually ships in 2 weeks.

Request a Bulk Order

assay

≥95% (LC/MS-UV)

form

solid

solubility

DMSO: 1 mg/mL

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O=C1C2=C(C=C(OCO3)C3=C2OC)OC(C4=CC=C(OC)C=C4)=C1OC

InChI

1S/C19H16O7/c1-21-11-6-4-10(5-7-11)16-19(23-3)15(20)14-12(26-16)8-13-17(18(14)22-2)25-9-24-13/h4-8H,9H2,1-3H3

InChI key

LWUQVHKGVXRUPD-UHFFFAOYSA-N

General description

Natural product derived from plant source.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service