Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00441

Sigma-Aldrich

Geniposidic acid

≥98% (HPLC)

Synonym(s):

Geniposidic acid, (1S,4aS,7aS)-1-(β-D-Glucopyranosyloxy)-1,4a,5,7a-tetrahydro-7-(hydroxymethyl)-cyclopenta[c]pyran-4-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22O10
CAS Number:
Molecular Weight:
374.34
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

biological source

plant (Rubiaceae gardenia)

assay

≥98% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

O[C@@H]1[C@@H](CO)O[C@@H](O[C@H]2[C@@](C(CO)=CC3)([H])[C@@]3([H])C(C(O)=O)=CO2)[C@H](O)[C@H]1O

InChI

1S/C16H22O10/c17-3-6-1-2-7-8(14(22)23)5-24-15(10(6)7)26-16-13(21)12(20)11(19)9(4-18)25-16/h1,5,7,9-13,15-21H,2-4H2,(H,22,23)/t7-,9-,10-,11-,12+,13-,15+,16+/m1/s1

InChI key

ZJDOESGVOWAULF-OGJQONSISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Geniposidic acid (GA) is a natural iridoid glucoside found in a variety of plants.

Biochem/physiol Actions

Geniposidic acid acts as an anti-inflammatory and anti-cancer agent, and has known effectiveness for jaundice and hepatic disorders. It is a moderator of COX inhibition, is a HSF1 inducer, and plays a role in collagen synthesis.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service