Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4701128

Sigma-Aldrich

Monoclonal Anti-CD94-APC antibody produced in mouse

clone HP-3D9

Synonym(s):

KLR-D1, KLRD1

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
$345.00

$345.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 TESTS
$345.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$345.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

mouse

conjugate

Allophycocyanin conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

HP-3D9, monoclonal

form

buffered aqueous solution

technique(s)

flow cytometry: 10 μL

accession no.

Q13241

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... KLRD1(3824)

Related Categories

General description

The mouse monoclonal antibody HP-3D9 recognizes CD94, a 70 kDa type II transmembrane glycoprotein expressed on NK cells, NK-T cells, and subsets of CD8+ T cells and gamma/delta T cells.

Immunogen

Cultured human NK cells

Physical form

Solution in phosphate buffered saline with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service