SAB1307045
ANTI-ZDHHC20(N-TERMINAL) antibody produced in rabbit
affinity isolated antibody, buffered aqueous solution
Synonym(s):
Probable palmitoyltransferase ZDHHC20, ZDHHC20, Zinc finger DHHC domain-containing protein 20
Select a Size
$496.00
Select a Size
About This Item
$496.00
biological source
rabbit
Quality Level
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
42278 Da
species reactivity
human
technique(s)
immunohistochemistry: 1:10-1:50
western blot: 1:1000
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... ZDHHC20(253832)
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service