Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S5151

Sigma-Aldrich

Ser-Gln-Asn-Tyr-Pro-Ile-Val

≥97% (HPLC)

Synonym(s):

HIV gag fragment 129-135

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H57N9O12
CAS Number:
Molecular Weight:
819.90
MDL number:
UNSPSC Code:
12352200

assay

≥97% (HPLC)

storage temp.

−20°C

Substrates

HIV-1 protease substrate, the minimum sequence to show such activity.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P L Darke et al.
The Journal of biological chemistry, 264(4), 2307-2312 (1989-02-05)
The protease of human immunodeficiency virus has been expressed in Escherichia coli and purified to apparent homogeneity. Immunoreactivity toward anti-protease peptide sera copurified with an activity that cleaved the structural polyprotein gag p55 and the peptide corresponding to the sequence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service