Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RAB0830

Sigma-Aldrich

Mouse Mme / Neprilysin ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 61 pg/mL
standard curve range: 61.44-15000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Mme(17380)

General description

The antibody pair provided in this kit recognizes mouse Neprilysin.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

8A - Combustible corrosive hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marianna E Jung et al.
Pharmacology, biochemistry, and behavior, 191, 172873-172873 (2020-02-28)
Many elderly American women use CNS depressant benzodiazepine (BZD) to ameliorate anxiety or insomnia. However, the chronic use of BZD (cBZD) is prevalent, causing adverse effects of BZD that include movement deficit. We previously reported that cBZD upregulates neurotoxic amyloid
Susanna Koskelainen et al.
Amyloid : the international journal of experimental and clinical investigation : the official journal of the International Society of Amyloidosis, 27(2), 81-88 (2019-12-10)
AGel amyloidosis is a dominantly inherited systemic amyloidosis caused by mutations p.D214N or p.D214Y resulting in gelsolin amyloid (AGel) formation. AGel accumulates extracellularly in many tissues and alongside elastic fibres. AGel deposition associates with elastic fibre degradation leading to severe
Aman Ullah Khan et al.
Microorganisms, 8(7) (2020-07-17)
Brucellosis is one of the most important worldwide zoonoses of many countries including Egypt. Camel brucellosis has not gained much attention in Egypt yet. This study is focused on the three governorates with the highest camel populations and the largest
Jiansheng Li et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 83, 153475-153475 (2021-02-06)
Bufei Yishen formula (BYF), a traditional Chinese medicine (TCM), is an effective therapeutic strategy for patients with chronic obstructive pulmonary disease (COPD). To evaluate the efficacy of BYF and investigate its therapeutic mechanisms. A total of 134 patients completed the
Lei Zuo et al.
Vaccines, 9(1) (2021-01-21)
Avian coronavirus infectious bronchitis virus (IBV) causes severe economic losses in the poultry industry, but its control is hampered by the continuous emergence of new genotypes and the lack of cross-protection among different IBV genotypes. We designed a new immunogen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service