Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0283

Sigma-Aldrich

PF-06745013

≥98% (HPLC)

Synonym(s):

2-{4-[6-Amino-5-(4-chloro-phenyl)-pyridin-3-yl]-phenoxy}-2-methyl-propionic acid, PF06745013

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H19ClN2O3
CAS Number:
Molecular Weight:
382.84
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

NC1=C(C2=CC=C(Cl)C=C2)C=C(C3=CC=C(OC(C)(C)C(O)=O)C=C3)C=N1

Biochem/physiol Actions

PF-06745013 is a potent and selective inhibitor of MAP4K4.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service