Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0101

Sigma-Aldrich

CP-226269

≥98% (HPLC)

Synonym(s):

5-Fluoro-2-{[4-(2-pyridinyl)-1-piperazinyl]methyl}-1H-indole

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$101.00

$101.00


Check Cart for Availability


Select a Size

Change View
5 MG
$101.00

About This Item

Empirical Formula (Hill Notation):
C18H19FN4
CAS Number:
Molecular Weight:
310.37
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

$101.00


Check Cart for Availability

assay

≥98% (HPLC)

form

powder

color

light tan

solubility

DMSO: >10 mg/mL

storage temp.

−20°C

SMILES string

Fc1ccc2[nH]c(CN3CCN(CC3)c4ccccn4)cc2c1

InChI

1S/C18H19FN4/c19-15-4-5-17-14(11-15)12-16(21-17)13-22-7-9-23(10-8-22)18-3-1-2-6-20-18/h1-6,11-12,21H,7-10,13H2

InChI key

PQOIDBZLMJMYCD-UHFFFAOYSA-N

Biochem/physiol Actions

CP-226269 is a D4 dopamine receptor agonist with EC50 of 32 nM.

Features and Benefits

This compound is featured on the Dopamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to dopamine receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service