Key Documents
P8427
Sigma-Aldrich
Pipette canisters
For 0.1 - 0.5 mL pipettes
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
size
51 mm × 51 mm × 406 mm
General description
Heavy gauge, satin finish stainless steel. Square cross-section permits easy autoclaving and storage. Silicone pads inside top and bottom minimize chipping of pipette tips.
Compatibility
For 0.1-0.5 mL pipettes
Choose from one of the most recent versions:
Certificates of Analysis (COA)