Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P6490

Sigma-Aldrich

PGP-4008

Synonym(s):

N-(1-Benzyl-2,3-dihydro-1H-pyrrolo[2,3-b]quinolin-4-yl)-2-phenylacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H23N3O
CAS Number:
Molecular Weight:
393.48
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

assay

≥90% (TLC)

Quality Level

form

powder

storage temp.

−20°C

SMILES string

O=C(Cc1ccccc1)Nc2c3CCN(Cc4ccccc4)c3nc5ccccc25

InChI

1S/C26H23N3O/c30-24(17-19-9-3-1-4-10-19)28-25-21-13-7-8-14-23(21)27-26-22(25)15-16-29(26)18-20-11-5-2-6-12-20/h1-14H,15-18H2,(H,27,28,30)

InChI key

HVIAKQBMYMKWII-UHFFFAOYSA-N

Application

Selectively inhibits P-glycoprotein.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C D Smith et al.
Oncology research, 12(5), 219-229 (2001-06-22)
Tumor cells often develop drug resistance through overexpression of membrane transport proteins that effectively efflux anticancer agents. The pharmacologies of the two best-studied transporters, P-glycoprotein (Pgp) and MRP1, are partially overlapping but distinct. To improve the therapeutic potential of drug

Articles

Protein-based drug transporters are expressed in Sf9 cells. Understanding the specific mechanisms of tumor cell transporters is an essential aspect of chemotherapeutic drug design.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service