Key Documents
P6241
Nunc® MaxiSorp 384 well plates
clear
About This Item
UNSPSC Code:
41100000
material
clear
sterility
non-sterile
feature
lid: no
packaging
case of 30 ea (internal packs of 10)
manufacturer/tradename
Nunc
maximum volume
120 μL
plate L × W
128 mm × 86 mm
suitability
suitable for (colorimetric assays, sample storage; luminescence and fluorescence assays, ELISA)
General description
MaxiSorp plates for ELISA
  • Same footprint as a 96 well plate
  • Working volume range of 10-100 μL
  • Compatible with robotic, automated readers and liquid handling systems
  • Assay miniaturization increases productivity and reduces cost
  • C-well for optical clarity and sample recovery
  • Reduced risk of well-to-well contamination
  • Optimizes robotic access to wells
  • White and black plates minimize crosstalk
  • Rounded square wells eliminate wicking (capillary action)
  • Flat bottom for optical reading
  • Optimized for scintillation counting
  • Black 384 well plate reduces background fluorescence and minimizes backlight scatter
  • White 384 well plate maximizes reflection of signal while maintaining low background
  • Clear 384 well plate for optical clarity for readthrough applications
Legal Information
MaxiSorp is a trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)