Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P3179

Sigma-Aldrich

Pyridinium tribromide

~95%

Synonym(s):

Pyridine hydrobromide perbromide, Pyridine perbromide hydrobromide, Pyridinium bromide perbromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N·HBr·Br2
CAS Number:
Molecular Weight:
319.82
Beilstein/REAXYS Number:
3690144
EC Number:
MDL number:
UNSPSC Code:
12352204

assay

~95%

storage temp.

2-8°C

SMILES string

Br[Br-]Br.C1=C[NH+]=CC=C1

InChI

1S/C5H5N.Br3/c1-2-4-6-5-3-1;1-3-2/h1-5H;/q;-1/p+1

InChI key

NSBNSZAXNUGWDJ-UHFFFAOYSA-O

Looking for similar products? Visit Product Comparison Guide

Application

Easily handled brominating agent

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service